Key Documents
Z552720
Sigma-Aldrich
Aldrich® Thermometer Adapter
straight, male joint: ST/NS 29/42, GL 14 thread
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
Joint
male joint: ST/NS29/42
fitting
GL 14 thread
Related Categories
General description
Has PTFE faced silicone washer and open-top cap.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC