Key Documents
BAH189620004
SP Bel-Art® Poxygrid® Extended Base Microliter Pipettor Rack
to hold, Gilson Pipetman
Synonym(s):
Gilson Pipetman pipettor rack, extended base pipette rack
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
feature
to hold Gilson Pipetman
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H18962-0004
L × W × H
9 in. (229 mm) × 7.875 in. (200 mm) × 6.75 in. (171 mm)
General description
Choose from 3 styles of epoxy-coated aluminum rack to hold Gilson Pipetman® pipettors.
Features and Benefits
  • Provides storage and easy access for up to 10 pipettors plus up to two Pipet-Aid® pipettes
  • H18962-0001 for desktop storage
  • H18962-0002 for wall mounting; has two holes for screw attachment to the wall and double faced tape (included)
  • H18962-0004 has extended base with raised lip to hold plastic tip boxes; suitable for wall mounting or for free standing on a bench
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Pipet-Aid is a registered trademark of Drummond Scientific Co.
Pipetman is a registered trademark of Gilson S.A.S. LLC
Poxygrid is a registered trademark of SP Scienceware, a division of SP