Skip to Content
Merck
All Photos(1)

Key Documents

916196

Sigma-Aldrich

Carbon coated Lithium Titanate (“LTO”) powder

battery grade

Synonym(s):

CC-LTO, CC-lithium titanium oxide, LTO-spinel with amorphous carbon, NANOMYTE® BE-10C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NA.23

Quality Level

grade

battery grade

form

powder

composition

Li4Ti5O12 and 1-3 wt% carbon

particle size

3-5 μm

application(s)

battery manufacturing

Application

Carbon coated LTO, battery grade is a class of anode material that is designed to improve rate performance for high-power Li-ion batteries and provides researcheres with an electronically conductive and effective battery material.

Legal Information

NANOMYTE is a registered trademark of NEI Corporation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morphologies and structures of carbon coated on Li4Ti5O12 and their effects on lithium storage performance.
Guo X, et al.
Electrochimica Acta, 130, 470-476 (2014)
Jean-Christophe Daigle et al.
Scientific reports, 9(1), 16871-16871 (2019-11-16)
Lithium titanium oxide (Li4Ti5O12)-based cells are a promising technology for ultra-fast charge-discharge and long life-cycle batteries. However, the surface reactivity of Li4Ti5O12 and lack of electronic conductivity still remains problematic. One of the approaches toward mitigating these problems is the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service