Skip to Content
Merck
All Photos(1)

Key Documents

244945

Sigma-Aldrich

cis-Dichlorobis(triphenylphosphine)platinum(II)

Synonym(s):

Pt(PPh3)2Cl2, cis-Bis(triphenylphosphine)platinum(II) dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)3P]2PtCl2
CAS Number:
Molecular Weight:
790.56
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

reaction suitability

core: platinum
reagent type: catalyst

mp

≥300 °C (lit.)

SMILES string

[Pt+2].P(c6ccccc6)(c5ccccc5)c4ccccc4.P(c3ccccc3)(c2ccccc2)c1ccccc1.[Cl-].[Cl-]

InChI

1S/2C18H15P.2ClH.Pt/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h2*1-15H;2*1H;/q;;;;+2/p-2

InChI key

XAFJSPPHVXDRIE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the platinum percentage in your product with catalog number 244945?

    1 answer
    1. The molecular weight of platinum is 195.084 g/mole. The molecular weight of this compound is 790.56 g/mole. The platinum is expected to be ~24.7% of this product. This is not routinely quality guaranteed as the titration result is based upon the % chlorine content.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service