Merck
Polyethyleneimine-poly(ethylene glycol)-star-copolymers as efficient and biodegradable vectors for mammalian cell transfection.
Journal of biomedical materials research. Part A (2013-07-31)
Katharina Ladewig, Zhi Ping Xu, Peter Gray, G Q Max Lu
RÉSUMÉ
High molecular weight (MW) polyethyleneimine (PEI) has been successfully used for the transfection of a broad variety of cell lines. In contrast to low MW PEI, which exhibits low transfection efficiencies but also low cytotoxicity, high MW PEI-mediated transfection achieves much higher efficiencies but at the cost of cell viability; therefore its use in commercial scale transfection and clinical application is limited. In this work we address this problem by constructing biodegradable high MW PEI mimics built from low MW PEI building blocks. The end-groups of small 5-arm star polyethylene glycol (PEG) prepolymers were decorated with linear oligo-ethyleneimine (OEI)/PEI arms of various MW via azomethine linkages. The resultant PEI-PEG-star-copolymers were investigated for their ability to complex plasmid DNA. Polymer/DNA complexes were characterized using techniques such as dynamic light scattering and transmission electron microscopy. Having established their cytotoxicity limits, they were tested as gene delivery vehicles for the transfection of suspension adapted Chinese hamster ovary (CHO-S) cells under serum-free conditions and adherent human embryonic kidney cells (HEK293T) in serum containing medium. Our PEI-PEG-star-copolymers showed a reduced cytotoxicity compared to high MW PEI while maintaining the ability to complex plasmid DNA and transfect mammalian cells, with significant transfection efficiencies. The effects of the optimum parameters on the transfection of mammalian cells using such novel polymers are discussed.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Branched PEI-g-PEG, PEG Mn 550
Sigma-Aldrich
Branched PEI-g-PEG, PEG Mn 5,000