Merck
  • Impaired M. tuberculosis Antigen-Specific IFN-γ Response without IL-17 Enhancement in Patients with Severe Cavitary Pulmonary Tuberculosis.
Impaired M. tuberculosis Antigen-Specific IFN-γ Response without IL-17 Enhancement in Patients with Severe Cavitary Pulmonary Tuberculosis.
PloS one (2015-05-29)
Lin Fan, Heping Xiao, Guangliang Mai, Bo Su, Joel Ernst, Zhongyi Hu
RÉSUMÉ
Th1 cells play an essential role in immune protection against tuberculosis. Th17 cells might be involved with immune pathology in active human tuberculosis (TB). The balance between Th1 and Th17 cells in patients with cavitary tuberculosis needs to be clarified which might help understanding the immunological basis of pathologic pathogenesis in TB. Initially treated pulmonary TB (PTB) patients with or without cavities were recruited before chemotherapy. We isolated peripheral blood mononuclear cells, stimulated with phytohemagglutinin (PHA), PPD, or ESAT-6 antigens, and assayed supernatant IFN-γ and IL-17 by ELISA after 24 or 72 hours incubation, respectively. Cells were also stained with antibodies to CD3, CD4, CD8, IFN-γ or IL-17 and the proportion of stained cells was measured by flow cytometry. We found wide variation of IFN-γ response in active PTB patients, but less subject-to-subject variation of IL-17 was observed as we previously reported. There were no significant differences in IFN-γ and IL-17 between cavitary and non-cavitary PTB; however, we found decreased IFN-γ secretion in severe cavitary PTB compared to mild lesion non-cavitary PTB (p < 0.05). We also found a decrease in the proportion of CD3+CD4+ T cells in the blood of severe cavitary PTB patients (p < 0.05). IL-17 seemed to have no association with the formation of cavities in active PTB from the study of PBMC. Impaired IFN-γ without IL-17 enhancement occurs in peripheral blood during severe cavitary PTB. Our results demonstrate that M. tuberculosis antigen-specific Th1 response is decreased when PTB lesions develop to severe cavities.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Interleukin-17A human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture