Merck
Mechanistic and structural studies on Rhodococcus ATCC 39484 nitrilase.
Biotechnology and applied biochemistry (1992-06-01)
D E Stevenson, R Feng, F Dumas, D Groleau, A Mihoc, A C Storer
RÉSUMÉ
Rhodococcus ATCC 39484 produced a nitrilase when induced with isovaleronitrile. The enzyme was obtainable pure in milligram amounts, had a subunit Mr of 40 kDa, and demonstrated a substrate-induced activation related to aggregation of subunits to form a 560-kDa complex. The enzyme had a broad substrate specificity, had a pH optimum of 7.5, was stable up to 40 degrees C, and had one disulfide bridge and two free cysteine residues, one of which appeared to be catalytically essential. The N-terminal sequence was determined and found to have 78.3% homology, in a 23-residue overlap, with Klebsiella ozaenae nitrilase. The enzyme was inhibited competitively by benzylamine and benzaldehyde and irreversibly by benzyl bromide. However, benzyl bromide was shown to be nonspecific, causing multiple alkylation. Acid quenching of enzyme-substrate mixtures allowed for the detection of covalent enzyme-substrate complexes using mass spectrometry. The covalent intermediate is suggested to be either a thioimidate or an acylenzyme and a reaction mechanism consistent with this observation and also the inhibitor results is proposed. The rate of breakdown of the covalent intermediates was found to be rate limiting even for substrates with undetectable rates of hydrolysis or those with very slow rates of intermediate formation. For phenylacetonitrile, a poor substrate, in addition to acid, approximately 2% of the product was the corresponding amide. This result suggests that a tetrahedral intermediate is formed which, for selected substrates, can break down anomalously to produce amide in place of the normal acid product. Under the conditions used in this study all other substrates tested were converted to acid.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Isovaleronitrile, 98%