Merck
[Cultivation of primary animal cells on microcarriers of various types].
Voprosy virusologii (1980-09-01)
M A Zaval'nyĭ, V P Grachev, T N Denisova, V L Popova, L L Mironova
RÉSUMÉ
Primary cells of chick embryos (CEC), new-born piglet kidneys (NPK), and green monkey kidney (GMK) obtained by the conventional method of tissue trypsinization were shown to grow satisfactorily on microcarriers of various types. CEC formed a confluent monolayer on Supperbead particles within 38--48 hours when seeded in a dose of 3 x 10(5) cells/ml and on DEAE-Sephadex A-50 within 60--72 hours when seeded in a dose of 4 x 10(5) cells/ml. In both instances, with microcarriers used in a concentration of 1 mg/ml the number of CEC by the end of the cultivation period reached 0.9 million cells/ml. Primary NPK and GMK cells formed monolayers not on all microcarrier particles, nevertheless the number of cells in both cultures increased 2--2 1/2-fold in 7 days. The optimal seed concentration of cells depended both on the quality of the primary cell suspension and on the microcarrier type. The monolayer of CEC cells formed on DEAE-Sephadex A-50 particles persisted for 7--14 days.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
DEAE–Sephadex®