Key Documents
Z740159
LifeSep magnetic separator unit
96F, for 96 well tray
Synonyme(s) :
cell sorting magnetic seperation unit
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NA.56
Caractéristiques
for 96 well tray
Fabricant/nom de marque
Dexter Magnetic Technologies 2501008-1
Description générale
Durable, easy-to-use and designed to accommodate industry standard microtitre plates. For use in cell sorting, RNA and DNA isolation and purification of biomolecules. Fast and easy aspiration is achieved with our Neodymium-Iron-Boron (Nd-Fe-B) magnet configuration.
  • Patented Nd-Fe-B magnet configuration for optimal results
  • Optimized field gradient within wells
  • Rapid separation of micro- and nanometer beads
  • Easy aspiration
  • Superior particle retention
  • Removable base for easy cleaning