Key Documents
GE10600069
Amersham Hybond® P Western blotting membranes, PVDF
pore size 0.45 μm, sheet W × L 140 mm × 160 mm, pkg of 25 ea
About This Item
Code UNSPSC :
41105339
Nomenclature NACRES :
NB.22
Conditionnement
pkg of 25 ea
Fabricant/nom de marque
Cytiva 10600069
Feuille, l × L
140 mm × 160 mm
Dimension de pores
0.45 μm pore size
Description générale
PVDF membranes have higher mechanical strength than NC membranes and, thus, are more suitable for Western blotting applications where stripping and reprobing is needed. In addition, PVDF membranes exhibit higher protein binding capacity, however, with a tendency for a higher background, compared with NC membranes.
Caractéristiques et avantages
  • Protein binding capacity over 200 µg/cm2 for easy signal detection
  • High protein retention even after harsh washing steps.
  • High protein capture during transfer minimizes sample loss.
  • Available in popular precut sizes for your application.
Stockage et stabilité
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Informations légales
Amersham is a trademark of Cytiva
Hybond is a registered trademark of Cytiva
Articles
This page describes troubleshooting strategies in the detection of GST-tagged proteins.