Key Documents
GBL206384
Grace Bio-Labs ProPlate MP microtiter plate superstructure
size 384 wells, black polystyrene, adhesive
Synonyme(s) :
high throughput robotic processing of microarrays, microarray system, microwell plate with permanently affixed well format
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.22
Matériaux
black polystyrene
Stérilité
non-sterile
Caractéristiques
wells 384
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Grace Bio-Labs 206384
Taille
384 wells
Description générale
Patented ProPlate® technology for high throughput robotic processing of microarrays.
The superstructures come in a permanently affixed well format--the ProPlate MP, or a removable well format--the FlexPlate MP. The ProPlate MP SBS compliant microtiter plate features a SecureSeal Plus adhesive bonding system applied to a bottomless 96-round well, black, polystyrene microtiter plate.
  • Guaranteed flat SBS compliant mircrotiter plate format which enables robotic processing of assays
  • Water-tight seal between superstructure and substrate
  • Compatible with glass or plastic substrates
  • Available in 96 round or square or 384 square well formats
Informations légales
FlexPlate MP is a trademark of Grace Bio-Labs, Inc.
ProPlate is a registered trademark of Grace Bio-Labs, Inc.
ProPlate MP is a trademark of Grace Bio-Labs, Inc.
SecureSeal is a trademark of Grace Bio-Labs, Inc.