Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Available to ship TODAYfromMILWAUKEE | $64.70 |
About This Item
NACRES:
NB.51
UNSPSC Code:
41122405
$64.70
Available to ship TODAYDetails
material
stainless steel
packaging
pack of 1 ea
L
4 in.
General description
with serrated tip
1 of 1
This Item | |||
|---|---|---|---|
| material stainless steel | material stainless steel | material stainless steel | material stainless steel |
| L 4 in. | L 8 in. | L 10 in. | L 4 3/4 in. |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Still not finding the right product?
Explore all of our products under Micro-dissecting forceps
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.