Key Documents
EP5702764000
Buckets for Eppendorf Rotor A-4-38
to hold, 16 x 125mm round bottom tubes
Synonyme(s) :
Round Buckets for Rotator A-4-38
About This Item
Code UNSPSC :
41103910
Nomenclature NACRES :
NB.15
Caractéristiques
to hold 16 x 125mm round bottom tubes
Conditionnement
pack of 4 ea
Fabricant/nom de marque
Eppendorf® 5702764000
Description générale
Centrifuge buckets to hold 16 x 125mm round bottom tubes. Compatible with Eppendorf rotor A-4-38.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG