Key Documents
Z531014
Sigma-Aldrich
Aldrich® Liebig condenser
Overall H 400 mm, joint: ST/NS 24/40
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS24/40
H totale
400 mm
Catégories apparentées
Description générale
Condensers have a standard taper female joint and a lower male drip tip-joint.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC