Key Documents
Z515647
Sigma-Aldrich
Aldrich® evaporating flask, pear-shaped
capacity 1,000 mL, joint: ST/NS 29/32, non-coated, heavy wall
Synonyme(s) :
bottom flask, flask round, round flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
VB.02
Matériaux
borosilicate glass
long
pear-shaped
single-neck
Joint
joint: ST/NS29/32
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
pack of 1
Fabricant/nom de marque
Büchi equivalent, 00435
Capacité
1,000 mL
Description générale
These pear-shaped evaporating flasks can be used as boiling type flasks for rotary evaporators.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC