Key Documents
Z408514
Sigma-Aldrich
Scienceware® wash bottle carrier
Holds 6 x 8/16 oz bottles
Synonyme(s) :
bottle carriers, bottle holder
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.17
Matériaux
polypropylene
Caractéristiques
Holds 6 x 8/16 oz bottles
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F11640-0001
L × l × H
286 mm × 194 mm × 225 mm
Description générale
Lightweight, PP carrier transports up to six 8 or 16 oz wash bottles. Autoclavable at 121°C. Bottles are not included.
Informations légales
Scienceware is a registered trademark of SP Scienceware, a division of SP