Key Documents
Z135674
Sigma-Aldrich
Polypropylene-coated support stand
With end rod
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.35
Taille de la base
8 in. × 12 in.
Diam. de la tige
1/2 in.
Description générale
No exposed metal parts. For use under severe corrosive conditions. Base is weighted. Rod screws into base with deep thread and tightening knob.