Key Documents
SYNF608350C
Synthware Buchner funnel
350 mL, porosity: coarse
Synonyme(s) :
Synthware F608350C, filtering funnel
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.51
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F608350C
Capacité
350 mL
Description générale
With sealed-in glass disc of the listed porosity. Not recommended for filtration where pressure would exceed 15psi. XC-extra coarse, C-Coarse, M-Medium, F-Fine porosity.
Informations légales
Synthware is a trademark of Kemtech America Inc.