Key Documents
SYNA721420
Synthware metering flow control adapter with PTFE stopcock
joint: ST/NS 14/20
Synonyme(s) :
Synthware A721420, flow control adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A721420
Catégories apparentées
Description générale
Provides on/off control with the main valve. Allows delivery of preset volumes of gases or liquids to reactions using the needle valve.
Informations légales
Synthware is a trademark of Kemtech America Inc.