Key Documents
BAF247390001
SP Bel-Art® Poxygrid Glove Dispenser
epoxy-coated (wire), to hold, Single Box Holder- holds 1x glove box
Synonyme(s) :
Glove box holder, glove dispenser
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.43
Matériaux
epoxy-coated (wire)
Caractéristiques
to hold Single Box Holder- holds 1x glove box
to hold Single Box Holder
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F24739-0001
H × L × width
8.25 in. (210 mm) × 5.5 in. (140 mm) × 4.25 in. (108 mm)
Description générale
Keep disposable gloves handy with these welded steel wire racks. Select rack to hold one, two or three boxes of disposable gloves, wipes or tissues.
Caractéristiques et avantages
  • Glove box drops into an individual retaining compartment and lifts out easily for replacement
  • Retaining compartment internal dimensions are 12.7cmW x 9.5cmD (5 x 3¾")
  • Designed for mounting to a vertical surface
  • Durable, blue epoxy coated racks are steam autoclavable at 121°C (250°F)
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP