Key Documents
932906
Sigma-Aldrich
Tweezer
EMS 4 WF
Synonyme(s) :
Gripper, Tweezer
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
NB.35
Description générale
EMS techware
Application
For the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips. Made from antimagnetic/anti-acid steel with smooth surface.
Tip Width: 12.0 mm. Depth: 9.0 mm. Tip Lip: E:2.5 mm. Offset: 4.0 mm.