ASTM D5501: GC Analysis of Ethanol Content in E85 Bioethanol on Petrocol® DH 150
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Petrocol DH 150, 150 m x 0.25 mm I.D., 1.0 μm (24155)
oven
60 °C (15 min), 30 °C/min to 250 °C (23 min)
inj. temp.
300 °C
detector
FID, 250 °C
carrier gas
helium, 24 cm/sec @ 60 °C
injection
0.5 μL, 150:1 split
liner
4 mm I.D., single taper
sample
E85 denatured fuel ethanol
Description
Legal Information
Petrocol is a registered trademark of Merck KGaA, Darmstadt, Germany