Merck
  • Unresolved recombination intermediates lead to ultra-fine anaphase bridges, chromosome breaks and aberrations.
Unresolved recombination intermediates lead to ultra-fine anaphase bridges, chromosome breaks and aberrations.
Nature cell biology (2017-12-20)
Ying Wai Chan, Kasper Fugger, Stephen C West
ABSTRACT
The resolution of joint molecules that link recombining sister chromatids is essential for chromosome segregation. Here, we determine the fate of unresolved recombination intermediates arising in cells lacking two nucleases required for resolution (GEN1 -/- knockout cells depleted of MUS81). We find that intermediates persist until mitosis and form a distinct class of anaphase bridges, which we term homologous recombination ultra-fine bridges (HR-UFBs). HR-UFBs are distinct from replication stress-associated UFBs, which arise at common fragile sites, and from centromeric UFBs. HR-UFBs are processed by BLM helicase to generate single-stranded RPA-coated bridges that are broken during mitosis. In the next cell cycle, DNA breaks activate the DNA damage checkpoint response, and chromosome fusions arise by non-homologous end joining. Consequently, the cells undergo cell cycle delay and massive cell death. These results lead us to present a model detailing how unresolved recombination intermediates can promote DNA damage and chromosomal instability.
Sigma-Aldrich
Anti-Chk1 antibody, Mouse monoclonal, ~2 mg/mL, clone DCS-310, purified from hybridoma cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human MRE11
Sigma-Aldrich
MISSION® esiRNA, targeting human BRCA2
Sigma-Aldrich
MISSION® esiRNA, targeting human DNA2
Sigma-Aldrich
MISSION® esiRNA, targeting human RECQL4
Sigma-Aldrich
MISSION® esiRNA, targeting human EXO1
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Anti-BRCA2 (Ab-1) Mouse mAb (2B), liquid, clone 2B, Calbiochem®
Sigma-Aldrich
MISSION® esiRNA, targeting human ERCC1
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting human WRN
Sigma-Aldrich
MISSION® esiRNA, targeting human MUS81
Sigma-Aldrich
Anti-PICH Antibody, clone 142-26-3, clone 142-26-3, from mouse
Sigma-Aldrich
Anti-Chk2 Antibody, clone 7, clone 7, Upstate®, from mouse