Merck
  • Use of a High-Throughput Phenotypic Screening Strategy to Identify Amplifiers, a Novel Pharmacological Class of Small Molecules That Exhibit Functional Synergy with Potentiators and Correctors.
Use of a High-Throughput Phenotypic Screening Strategy to Identify Amplifiers, a Novel Pharmacological Class of Small Molecules That Exhibit Functional Synergy with Potentiators and Correctors.
SLAS discovery : advancing life sciences R & D (2017-09-13)
Kenneth A Giuliano, Shinichiro Wachi, Lawrence Drew, Danijela Dukovski, Olivia Green, Cecilia Bastos, Matthew D Cullen, Sheila Hauck, Bradley D Tait, Benito Munoz, Po-Shun Lee, John Preston Miller
ABSTRACT
Cystic fibrosis (CF) is a lethal genetic disorder caused by mutation of the cystic fibrosis transmembrane conductance regulator (CFTR) gene. Despite recent groundbreaking approval of genotype-specific small-molecule drugs, a significant portion of CF patients still lack effective therapeutic options that address the underlying cause of the disease. Through a phenotypic high-throughput screen of approximately 54,000 small molecules, we identified a novel class of CFTR modulators called amplifiers. The identified compound, the characteristics of which are represented here by PTI-CH, selectively increases the expression of immature CFTR protein across different CFTR mutations, including F508del-CFTR, by targeting the inefficiencies of early CFTR biosynthesis. PTI-CH also augments the activity of other CFTR modulators and was found to possess novel characteristics that distinguish it from CFTR potentiator and corrector moieties. The PTI-CH-mediated increase in F508del-CFTR did not elicit cytosolic or endoplasmic reticulum-associated cellular stress responses. Based on these data, amplifiers represent a promising new class of CFTR modulators for the treatment of CF that can be used synergistically with other CFTR modulators.
Product Description
SAFC
Nutrient Mixture F-12 Ham, powder, with L-glutamine and 0.863 mg/L zinc sulfate, without sodium bicarbonate, Coon′s Modification, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Pricing and availability is not currently available.