Merck
Validation of a laboratory-developed test of human sperm capacitation.
Molecular reproduction and development (2017-04-19)
Melissa A Moody, Cristina Cardona, Alana J Simpson, T Timothy Smith, Alexander J Travis, G Charles Ostermeier
ABSTRACT
Sperm must undergo capacitation to become fertilization competent. Here we validated that monosialotetrahexosylganglioside (G
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
CTB, ≥98% (HPLC)