Merck
  • Exploring Step-by-Step Assembly of Nanoparticle:Cytochrome Biohybrid Photoanodes.
Exploring Step-by-Step Assembly of Nanoparticle:Cytochrome Biohybrid Photoanodes.
ChemElectroChem (2017-09-19)
Ee Taek Hwang, Katherine L Orchard, Daisuke Hojo, Joseph Beton, Colin W J Lockwood, Tadafumi Adschiri, Julea N Butt, Erwin Reisner, Lars J C Jeuken
ABSTRACT
Coupling light-harvesting semiconducting nanoparticles (NPs) with redox enzymes has been shown to create artificial photosynthetic systems that hold promise for the synthesis of solar fuels. High quantum yields require efficient electron transfer from the nanoparticle to the redox protein, a property that can be difficult to control. Here, we have compared binding and electron transfer between dye-sensitized TiO
Sigma-Aldrich
6-Amino-1-hexanethiol hydrochloride
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
Sulfur, 99.998% trace metals basis
Sigma-Aldrich
8-Amino-1-octanethiol hydrochloride, 95%
Sigma-Aldrich
2-(Dimethylamino)ethanethiol hydrochloride, 95%
Sigma-Aldrich
8-Mercapto-1-octanol, 98%