Merck
  • Chemical Modulation of Protein O-GlcNAcylation via OGT Inhibition Promotes Human Neural Cell Differentiation.
Chemical Modulation of Protein O-GlcNAcylation via OGT Inhibition Promotes Human Neural Cell Differentiation.
ACS chemical biology (2017-05-26)
Lissette M Andres, Ian W Blong, Angela C Evans, Neil G Rumachik, Teppei Yamaguchi, Nam D Pham, Pamela Thompson, Jennifer J Kohler, Carolyn R Bertozzi
ABSTRACT
The enzymes that determine protein O-GlcNAcylation, O-GlcNAc transferase (OGT) and O-GlcNAcase (OGA), act on key transcriptional and epigenetic regulators, and both are abundantly expressed in the brain. However, little is known about how alterations in O-GlcNAc cycling affect human embryonic stem cell (hESC) neural differentiation. Here, we studied the effects of perturbing O-GlcNAcylation during neural induction of hESCs using the metabolic inhibitor of OGT, peracetylated 5-thio-N-acetylglucosamine (Ac4-5SGlcNAc). Treatment of hESCs with Ac4-5SGlcNAc during induction limited protein O-GlcNAcylation and also caused a dramatic decrease in global levels of UDP-GlcNAc. Concomitantly, a subpopulation of neural progenitor cells (NPCs) acquired an immature neuronal morphology and expressed early neuronal markers such as β-III tubulin (TUJ1) and microtubule associated protein 2 (MAP2), phenotypes that took longer to manifest in the absence of OGT inhibition. These data suggest that chemical inhibition of OGT and perturbation of protein O-GlcNAcylation accelerate the differentiation of hESCs along the neuronal lineage, thus providing further insight into the dynamic molecular mechanisms involved in neuronal development.
Sigma-Aldrich
Milli-Mark® Anti-MAP2-PE Antibody, clone AP20, clone AP20, Milli-Mark®, from mouse
Sigma-Aldrich
Anti-MAP2 Antibody, clone AP20, clone AP20, Chemicon®, from mouse
Sigma-Aldrich
IgG, Mouse
Sigma-Aldrich
IgG, Rabbit, Rabbit IgG Polyclonal Antibody validated for use in ELISA.
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-Tubulin Antibody, beta III isoform, CT, clone TU-20 (Similar to TUJ1), ascites fluid, clone TU-20 (Similar to TUJ1), Chemicon®
Sigma-Aldrich
MTT Cell Growth Assay Kit, MTT Cell Growth Assay is a colorimetric assay that can be used for either proliferation or complement-mediated cytotoxicity assays.