Merck
  • (S)-5-ethynyl-anabasine, a novel compound, is a more potent agonist than other nicotine alkaloids on the nematode Asu-ACR-16 receptor.
(S)-5-ethynyl-anabasine, a novel compound, is a more potent agonist than other nicotine alkaloids on the nematode Asu-ACR-16 receptor.
International journal for parasitology. Drugs and drug resistance (2016-12-30)
Fudan Zheng, Xiangwei Du, Tsung-Han Chou, Alan P Robertson, Edward W Yu, Brett VanVeller, Richard J Martin
ABSTRACT
Nematode parasites infect ∼2 billion people world-wide. Infections are treated and prevented by anthelmintic drugs, some of which act on nicotinic acetylcholine receptors (nAChRs). There is an unmet need for novel therapeutic agents because of concerns about the development of resistance. We have selected Asu-ACR-16 from a significant nematode parasite genus, Ascaris suum, as a pharmaceutical target and nicotine as our basic moiety (EC
Sigma-Aldrich
Acetylcholine bromide, ≥99%
Sigma-Aldrich
Ethynyltrimethylsilane, 98%
Sigma-Aldrich
4,4′-Di-tert-butyl-2,2′-dipyridyl, 98%