Merck
  • Proteomics method to quantify the percentage of cow, goat, and sheep milks in raw materials for dairy products.
Proteomics method to quantify the percentage of cow, goat, and sheep milks in raw materials for dairy products.
Journal of dairy science (2016-10-25)
Q Chen, X Ke, J S Zhang, S Y Lai, F Fang, W M Mo, Y P Ren
ABSTRACT
Fraud in milk and dairy products occurs when cow milk is added to sheep and goat milk for economic reasons. No reliable, selective, and sensitive method exists for quantifying the milk percentage of different species. This work reports the development and validation of a proteomics-based method for the qualitative detection and quantitative determination of cow, sheep, and goat milks in the raw materials used for dairy products. β-Lactoglobulin was selected as the protein marker because it is a major protein in milk and whey powder. The tryptic peptides LSFNPTQLEEQCHI and LAFNPTQLEGQCHV were used as signature peptides for cow milk and for sheep and goat milks, respectively. The winged peptides LKALPMHIRLSFNPTQL*EEQCHI* and LKALPMHIRLAFNPTQL*EGQCHV* were designed and synthesized as internal standards. Validation of the method showed that it has good sensitivity, specificity, reproducibility, precision, and accuracy. This method is easily applicable in routine laboratory analysis without intensive proteomics background.
Sigma-Aldrich
Fmoc-Val-OH-13C5,15N, 98 atom % 15N, 98 atom % 13C, 99% (CP)
Sigma-Aldrich
Fmoc-Leu-OH-13C6,15N, 98 atom % 15N, 98 atom % 13C, 99% (CP)
Sigma-Aldrich
Indole-2-acetic acid
Sigma-Aldrich
Fmoc-Ile-OH-13C6,15N, 98 atom % 13C, 98 atom % 15N, 98% (CP)