Merck
  • SUMO2 overexpression enhances the generation and function of interleukin-17-producing CD8⁺ T cells in mice.
SUMO2 overexpression enhances the generation and function of interleukin-17-producing CD8⁺ T cells in mice.
Cellular signalling (2015-03-13)
Tae Joon Won, Yun-Jung Lee, Kyeong Eun Hyung, Eunyoung Yang, Uy Dong Sohn, Hae Young Min, Do Ik Lee, So-Young Park, Kwang Woo Hwang
ABSTRACT
Small ubiquitin-like modifier (SUMO) 2 is a small protein that controls the activity and stability of other proteins by SUMOylation. In this study, T cell-specific SUMO2 overexpressing transgenic mice were generated to study the effect of SUMO2 on T lymphocytes. SUMO2 overexpression promoted differentiation of interleukin (IL)-17-producing CD8(+) T cells, and significantly suppressed the growth of EL4 tumor cells in vivo. Moreover, the tumor tissue from SUMO2-overexpressing mice had higher interferon (IFN)-γ and granzyme B mRNA levels. Although SUMO2 overexpression did not increase IFN-γ or granzyme B production in cytotoxic T lymphocytes, IL-12 treatment restored and increased IFN-γ secretion in IL-17-producing CD8(+) T cells. SUMO2 overexpression also increased gene expression of chemokines, CCL4, and CXCL10, which attract cytotoxic T lymphocytes to tumor tissues. Additionally, SUMO2-overexpressing T cells exhibited increased STAT3 phosphorylation, implying a SUMO2 target which up-regulates STAT3 activity governing IL-17A-producing CD8(+) T cell differentiation and antitumor immune responses.
Sigma-Aldrich
Interleukin-2, human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
IL-2 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Roche
Interleukin-2, mouse (mIL-2), recombinant (E. coli)
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, carrier free
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder