Merck
  • An official American Thoracic Society statement: diagnosis and management of beryllium sensitivity and chronic beryllium disease.
An official American Thoracic Society statement: diagnosis and management of beryllium sensitivity and chronic beryllium disease.
American journal of respiratory and critical care medicine (2014-11-15)
John R Balmes, Jerrold L Abraham, Raed A Dweik, Elizabeth Fireman, Andrew P Fontenot, Lisa A Maier, Joachim Muller-Quernheim, Gaston Ostiguy, Lewis D Pepper, Cesare Saltini, Christine R Schuler, Tim K Takaro, Paul F Wambach
ABSTRACT
Beryllium continues to have a wide range of industrial applications. Exposure to beryllium can lead to sensitization (BeS) and chronic beryllium disease (CBD). The purpose of this statement is to increase awareness and knowledge about beryllium exposure, BeS, and CBD. Evidence was identified by a search of MEDLINE. The committee then summarized the evidence, drew conclusions, and described their approach to diagnosis and management. The beryllium lymphocyte proliferation test is the cornerstone of both medical surveillance and the diagnosis of BeS and CBD. A confirmed abnormal beryllium lymphocyte proliferation test without evidence of lung disease is diagnostic of BeS. BeS with evidence of a granulomatous inflammatory response in the lung is diagnostic of CBD. The determinants of progression from BeS to CBD are uncertain, but higher exposures and the presence of a genetic variant in the HLA-DP β chain appear to increase the risk. Periodic evaluation of affected individuals can detect disease progression (from BeS to CBD, or from mild CBD to more severe CBD). Corticosteroid therapy is typically administered when a patient with CBD exhibits evidence of significant lung function abnormality or decline. Medical surveillance in workplaces that use beryllium-containing materials can identify individuals with BeS and at-risk groups of workers, which can help prioritize efforts to reduce inhalational and dermal exposures.
Product Description
Beryllium, wire reel, 0.1m, diameter 0.25mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.1m, diameter 0.6mm, annealed and clean, 99%
Beryllium, wire reel, 5m, diameter 0.05mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.2m, diameter 0.6mm, annealed and clean, 99%
Beryllium, wire reel, 0.2m, diameter 0.85mm, annealed and clean, 99%
Beryllium, wire reel, 0.2m, diameter 0.25mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.5m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 1m, diameter 0.05mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.3m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.1m, diameter 0.85mm, annealed and clean, 99%
Beryllium, wire reel, 0.1m, diameter 0.05mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.05m, diameter 0.5mm, annealed and clean, 99%
Beryllium, wire reel, 0.1m, diameter 0.125mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.2m, diameter 0.125mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.05m, diameter 0.6mm, annealed and clean, 99%
Beryllium, wire reel, 0.2m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.1m, diameter 0.75mm, annealed and clean, 99%
Beryllium, wire reel, 0.05m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.1m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.05m, diameter 0.75mm, annealed and clean, 99%
Beryllium, wire reel, 0.1m, diameter 0.5mm, annealed and clean, 99%
Beryllium, wire reel, 0.2m, diameter 0.5mm, annealed and clean, 99%
Beryllium, wire reel, 0.05m, diameter 0.25mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.05m, diameter 0.85mm, annealed and clean, 99%
Beryllium, wire reel, 0.2m, diameter 0.75mm, annealed and clean, 99%
Beryllium, wire reel, 0.5m, diameter 0.05mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.5m, diameter 0.125mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.5m, diameter 0.25mm, annealed and clean, 99.7%
Beryllium, wire reel, 1m, diameter 0.25mm, annealed and clean, 99.7%
Beryllium, wire reel, 1m, diameter 0.125mm, annealed and clean, 99.7%