Merck
  • Ber-MAC3: new monoclonal antibody that defines human monocyte/macrophage differentiation antigen.
Ber-MAC3: new monoclonal antibody that defines human monocyte/macrophage differentiation antigen.
Journal of clinical pathology (1991-11-01)
E Backé, R Schwarting, J Gerdes, M Ernst, H Stein
ABSTRACT
A new monoclonal antibody Ber-MAC3 is reported. It recognises a formol sensitive epitope of a not yet clustered monocyte/macrophage specific 140 kilodalton glycoprotein that is expressed on the cell surface and in the cytoplasm. In 30 cases of acute and chronic leukaemia, Ber-MAC3 staining was restricted to 15 myeloid leukaemias of M4 and M5 types. The tumour cells of two cases of true histiocytic malignancies were Ber-MAC3 positive, whereas those of all 280 malignancies of lymphocytic origin were negative. The latter included 52 cases of Hodgkin's disease and 41 cases of Ki-1 positive anaplastic large cell lymphomas which had previously been classified as true histiocytic lymphomas. Ber-MAC3 therefore seems to be of considerable value for selective identification of monocytes and macrophages at a certain stage of differentiation and seems to be suitable for diagnosing myelomonocytic or monocytic leukaemia and neoplasms of true histiocytic origin.
Sigma-Aldrich
CD163 (MRQ-26) Mouse Monoclonal Antibody