Merck
  • The toxic effect of oxytetracycline and trimethoprim in the aquatic environment.
The toxic effect of oxytetracycline and trimethoprim in the aquatic environment.
Chemosphere (2014-04-08)
Boris Kolar, Lovro Arnuš, Bogdana Jeretin, Aleš Gutmaher, Damjana Drobne, Mojca Kos Durjava
ABSTRACT
The objective of our study was the investigation of the toxic properties of two antimicrobial drugs: oxytetracycline (OTC) and trimethoprim (TMP) in the aquatic environment. The toxic effects were tested according to the OECD guidelines for the testing of chemicals, on the cyanobacteria Anabaena flos-aque, on the alga Pseudokirchneriella subcapitata, on the daphnid Daphnia magna as well as on the activated sludge. We discussed the short term and long term results of tests on cyanobacteria and microalgae. Both experiments were concluded in 72h allowing direct comparison of sensitivity of the two tested species. The results of our study showed toxic effect in the same range for both groups. In the test on the toxicity of OTC to P. subcapitata we obtained the 72hErC50 of 1.04mgL(-1) (72hErC10 0.47mgL(-1)) which are lower in comparison to the results on the toxicity to A. flos-aque of ErC50 of 2.7mgL(-1) (72hErC101.5mgL(-1)). TMP is less toxic to both photosynthetic plankton species. Similar to the test results on OTC, the P. subcapitata is more sensitive to TMP (ErC50129mgL(-1); ErC1065mgL(-1)) than A. flos-aque (72hErC50253mgL(-1); 72hErC1026mgL(-1)). OTC is toxic to the activated sludge (3hEC50 17.9mgL(-1)), while the calculated 3hEC50 value for TMP exceeded solubility for the compound. In comparison to other species, both tested antimicrobials showed low toxicity to daphnids.
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Supelco
Trimethoprim, VETRANAL®, analytical standard
Supelco
Oxytetracycline hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Oxytetracycline dihydrate, Assay (anhydrous basis) 95.0%-102.0%
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Oxytetracycline hydrochloride
USP
Oxytetracycline hydrochloride, United States Pharmacopeia (USP) Reference Standard
USP
Trimethoprim, United States Pharmacopeia (USP) Reference Standard
Supelco
Oxytetracycline Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Oxytetracycline hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Oxytetracycline, United States Pharmacopeia (USP) Reference Standard
Trimethoprim, European Pharmacopoeia (EP) Reference Standard
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Oxytetra Selective Supplement, suitable for microbiology