Merck
  • 2'-OMe-phosphorodithioate-modified siRNAs show increased loading into the RISC complex and enhanced anti-tumour activity.
2'-OMe-phosphorodithioate-modified siRNAs show increased loading into the RISC complex and enhanced anti-tumour activity.
Nature communications (2014-03-13)
Sherry Y Wu, Xianbin Yang, Kshipra M Gharpure, Hiroto Hatakeyama, Martin Egli, Michael H McGuire, Archana S Nagaraja, Takahito M Miyake, Rajesha Rupaimoole, Chad V Pecot, Morgan Taylor, Sunila Pradeep, Malgorzata Sierant, Cristian Rodriguez-Aguayo, Hyun J Choi, Rebecca A Previs, Guillermo N Armaiz-Pena, Li Huang, Carlos Martinez, Tom Hassell, Cristina Ivan, Vasudha Sehgal, Richa Singhania, Hee-Dong Han, Chang Su, Ji Hoon Kim, Heather J Dalton, Chandra Kovvali, Khandan Keyomarsi, Nigel A J McMillan, Willem W Overwijk, Jinsong Liu, Ju-Seog Lee, Keith A Baggerly, Gabriel Lopez-Berestein, Prahlad T Ram, Barbara Nawrot, Anil K Sood
ABSTRACT
Improving small interfering RNA (siRNA) efficacy in target cell populations remains a challenge to its clinical implementation. Here, we report a chemical modification, consisting of phosphorodithioate (PS2) and 2'-O-Methyl (2'-OMe) MePS2 on one nucleotide that significantly enhances potency and resistance to degradation for various siRNAs. We find enhanced potency stems from an unforeseen increase in siRNA loading to the RNA-induced silencing complex, likely due to the unique interaction mediated by 2'-OMe and PS2. We demonstrate the therapeutic utility of MePS2 siRNAs in chemoresistant ovarian cancer mouse models via targeting GRAM domain containing 1B (GRAMD1B), a protein involved in chemoresistance. GRAMD1B silencing is achieved in tumours following MePS2-modified siRNA treatment, leading to a synergistic anti-tumour effect in combination with paclitaxel. Given the previously limited success in enhancing siRNA potency with chemically modified siRNAs, our findings represent an important advance in siRNA design with the potential for application in numerous cancer types.
Sigma-Aldrich
Anti-Eck/EphA2 Antibody, clone D7, clone D7, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid