Merck
  • Pharmacokinetic comparison between Conpremin (Premarin) and a generic preparation of conjugated estrogens.
Pharmacokinetic comparison between Conpremin (Premarin) and a generic preparation of conjugated estrogens.
Climacteric : the journal of the International Menopause Society (2002-03-23)
E Arteaga, P Villaseca
ABSTRACT
To determine the relative bioavailability of the estrogenic components of a generic brand of conjugated estrogens marketed in Chile in comparison to that of Conpremin (Premarin in the United States). A randomized cross-over study was conducted on 16 healthy postmenopausal women receiving single oral doses of either two Conpremin 0.625-mg tablets or two 0.625-mg tablets of the generic brand, with a 14-day wash-out interval between doses. A gas chromatography tandem mass spectrometry assay was used to determine estrogen components. The peak plasma concentrations of unconjugated and total estrone and equilin, unconjugated 17 beta-dihydroequilin and 17 beta-estradiol were higher and occurred earlier with the generic conjugated estrogens than with Conpremin. The 90% confidence limits for both variables lay outside the accepted bioequivalence limits of 80-125%. Additionally, no measurable plasma concentration of unconjugated delta 8,9-dehydroestrone or 17 beta-delta 8,9-dehydroestradiol was seen after administration of the generic conjugated estrogens. These pharmacokinetic results indicate that the generic tablets do not have the modified-release characteristics of Conpremin tablets. In addition, the absence of delta 8,9-dehydroestrone and 17 beta-delta 8,9-dehydroestradiol in the plasma indicates that the generic form is not compositionally equivalent to Conpremin.
Product Description
17α-Dihydroequilin, European Pharmacopoeia (EP) Reference Standard