Merck
  • Pulmonary embolism associated with protein C deficiency and abuse of anabolic-androgen steroids.
Pulmonary embolism associated with protein C deficiency and abuse of anabolic-androgen steroids.
Clinical and applied thrombosis/hemostasis : official journal of the International Academy of Clinical and Applied Thrombosis/Hemostasis (2008-11-04)
Alaa Alhadad, Stefan Acosta, Latif Sarabi, Tilo Kölbel
ABSTRACT
We present the case of a 19-year-old male athlete with protein C deficiency who developed proximal deep venous thrombosis and pulmonary embolism while abusing anabolic-androgenic steroids. Anabolic-androgenic steroids have been reported to have anticoagulatory and profibrinolytic effects in patients with protein C deficiency. Despite these antithrombotic effects, the patient developed repeated venous thromboembolism during treatment with low-molecular-weight heparin. The net effect of anabolic-androgenic steroids on the haemostatic system may change from antithrombotic to prothrombotic in male abusers of anabolic steroids with protein C deficiency.
Product Description
Supelco
17β-Hydroxy-17-methylandrosta-1,4-dien-3-one, analytical standard
Supelco
Methandienone solution, 1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®