Merck
  • Cytochrome P450 initiates degradation of cis-dichloroethene by Polaromonas sp. strain JS666.
Cytochrome P450 initiates degradation of cis-dichloroethene by Polaromonas sp. strain JS666.
Applied and environmental microbiology (2013-01-29)
Shirley F Nishino, Kwanghee A Shin, James M Gossett, Jim C Spain
ABSTRACT
Polaromonas sp. strain JS666 grows on cis-1,2-dichoroethene (cDCE) as the sole carbon and energy source under aerobic conditions, but the degradation mechanism and the enzymes involved are unknown. In this study, we established the complete pathway for cDCE degradation through heterologous gene expression, inhibition studies, enzyme assays, and analysis of intermediates. Several lines of evidence indicate that a cytochrome P450 monooxygenase catalyzes the initial step of cDCE degradation. Both the transient accumulation of dichloroacetaldehyde in cDCE-degrading cultures and dichloroacetaldehyde dehydrogenase activities in cell extracts of JS666 support a pathway for degradation of cDCE through dichloroacetaldehyde. The mechanism minimizes the formation of cDCE epoxide. The molecular phylogeny of the cytochrome P450 gene and the organization of neighboring genes suggest that the cDCE degradation pathway recently evolved in a progenitor capable of degrading 1,2-dichloroethane either by the recruitment of the cytochrome P450 monooxygenase gene from an alkane catabolic pathway or by selection for variants of the P450 in a preexisting 1,2-dichloroethane catabolic pathway. The results presented here add yet another role to the broad array of productive reactions catalyzed by cytochrome P450 enzymes.
Product Description
Supelco
cis-1,2-Dichloroethene, analytical standard
Sigma-Aldrich
cis-1,2-Dichloroethylene, 97%
Supelco
trans-1,2-Dichloroethylene, analytical standard
Sigma-Aldrich
trans-1,2-Dichloroethylene, 98%
Sigma-Aldrich
1,2-Dichloroethylene, mixture of cis and trans, 98%
Supelco
cis-1,2-Dichloroethene solution, certified reference material, 2000 μg/mL in methanol