Merck
Diagnosis and management of alcoholic hepatitis.
Clinics in liver disease (2012-10-30)
Umair Sohail, Sanjaya K Satapathy
ABSTRACT
Alcoholic hepatitis is a form of severe, cholestatic liver disease that results from consumption of large amount of alcohol during a sustained period of time in a subset of alcoholics. Symptoms could be mild and nonspecific to more severe. The diagnosis of alcoholic hepatitis can be made with a thorough history, physical examination, and review of laboratory results. Liver biopsy is confirmatory but generally not indicated for the diagnosis. Abstinence is the key form of therapeutic intervention. Despite variable results in clinical trials, corticosteroids and pentoxifylline seem to provide moderate survival benefit. Liver transplantation in acute alcoholic hepatitis is contentious.
Product Description
Pentoxifylline, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pentoxifylline, solid