Merck
  • Synthesis and biological evaluation of phenyl substituted 1H-1,2,4-triazoles as non-steroidal inhibitors of 17β-hydroxysteroid dehydrogenase type 2.
Synthesis and biological evaluation of phenyl substituted 1H-1,2,4-triazoles as non-steroidal inhibitors of 17β-hydroxysteroid dehydrogenase type 2.
Archiv der Pharmazie (2012-04-26)
Yaseen A Al-Soud, Sandrine Marchais-Oberwinkler, Martin Frotscher, Rolf W Hartmann
ABSTRACT
A series of disubstituted-1H-1,2,4-triazole derivatives was synthesized with the aim of developing new non-steroidal inhibitors of 17β-hydroxysteroid dehydrogenase type 2 (17βHSD2) - a novel and attractive target for the treatment of osteoporosis. 17βHSD2 catalyzes the oxidation of the highly active estrogen 17β-estradiol (E2) and androgen testosterone (T) into the weak estrone and androstenedione, respectively. Inhibition of this enzyme will locally in the bone lead to an increase in E2 and T levels, two key players in the maintenance of the balance between bone resorption and bone formation. In this study, a new class of 17βHSD2 inhibitors with a 1H-1,2,4-triazole scaffold was identified; the three best compounds 8b, 8f, and 13a showed moderate 17βHSD2 inhibitory activity and a good selectivity toward 17βHSD1. They could be a useful tool to map the unexplored enzyme active site.
Sigma-Aldrich
1,2,4-Triazole, 98%