Merck
  • Enhanced 2.0 microm emission and gain coefficient of transparent glass ceramic containing BaF2: Ho3+,Tm3+ nanocrystals.
Enhanced 2.0 microm emission and gain coefficient of transparent glass ceramic containing BaF2: Ho3+,Tm3+ nanocrystals.
Optics express (2009-12-10)
W J Zhang, Q Y Zhang, Q J Chen, Q Qian, Z M Yang, J R Qiu, P Huang, Y S Wang
ABSTRACT
Transparent glass ceramic containing BaF(2):Ho(3+),Tm(3+) nanocrystals has been prepared by melt quenching and subsequent thermal treatment. The precipitation of BaF(2) nanocrystals was confirmed by X-ray diffraction and high-resolution transmission electron microscopy. Intense 2.0 microm fluorescence originating from Ho(3+): (5)I(7) --> (5)I(8) transition was achieved upon excitation with 808 nm laser diode. A large ratio of forward Tm(3+) --> Ho(3+) energy transfer constant to that of backward process indicated high efficient energy transfer from Tm(3+)((3)F(4)) to Ho(3+)((5)I(7)), benefited from the reduced ionic distances of Tm(3+)-Tm(3+) and Tm(3+)-Ho(3+) pairs and low phonon energy environment with the incorporation of rare-earth ions into the precipitated BaF(2) nanocrystals. The results indicate that glass ceramic is a promising candidate material for 2.0 microm laser.
Sigma-Aldrich
Barium fluoride, powder, 98%
Sigma-Aldrich
Barium fluoride, precipitated, 99.95% trace metals basis
Sigma-Aldrich
Barium fluoride, 99.99% trace metals basis