Merck
  • Palladium-catalyzed intramolecular direct arylation of benzoic acids by tandem decarboxylation/C-H activation.
Palladium-catalyzed intramolecular direct arylation of benzoic acids by tandem decarboxylation/C-H activation.
Journal of the American Chemical Society (2009-03-26)
Congyang Wang, Isabel Piel, Frank Glorius
ABSTRACT
A novel protocol for the palladium-catalyzed intramolecular direct arylation of benzoic acids is reported, which combines decarboxylation and C-H activation into one efficient process. With the optimized catalyst system of palladium trifluoroacetate, silver carbonate, and the proper solvent mixture (5% DMSO/dioxane), dibenzofuran derivatives are formed in excellent yield and selectivity.
Sigma-Aldrich
Silver carbonate on Celite®, extent of labeling: ~50 wt. % loading
Sigma-Aldrich
Silver carbonate, purum p.a., ≥99.0% (AT)
Sigma-Aldrich
Silver carbonate, 99%