Merck
  • DNA-protein cross-link formation in Burkitt lymphoma cells cultured with benzaldehyde and the sedative paraldehyde.
DNA-protein cross-link formation in Burkitt lymphoma cells cultured with benzaldehyde and the sedative paraldehyde.
Drug and chemical toxicology (2007-03-17)
J R Kuykendall, E J Jarvi, B L Finley, D J Paustenbach
ABSTRACT
Exposure to aldehydes represents potential risks to human and animal health. Cyclic aldehydes such as benzaldehyde, 2-furaldehyde, and paraldehyde were found to induce formation of stable DNA-protein cross-links (DPXs) in cultured human lymphoma cells. A relationship between increased cytotoxicity and DPX formation was observed with each aldehyde. Paraldehyde is a sedative drug used predominately in treatment of ethanol withdrawal. Paraldehyde was the most potent cross-linking aldehyde studied, yet least cytotoxic. Although DPX formation by aliphatic aldehydes is well-known, this study confirms the potential for cyclic aldehydes to cause formation of DPXs in cultured cells at therapeutically relevant doses.
Sigma-Aldrich
Paraldehyde, ≥97.0% (GC)
Sigma-Aldrich
Paraldehyde, ≥97%
Sigma-Aldrich
Paraldehyde, Arxada quality, ≥99.5% (GC)