Merck
  • Structural studies of a two-domain chitinase from Streptomyces griseus HUT6037.
Structural studies of a two-domain chitinase from Streptomyces griseus HUT6037.
Journal of molecular biology (2006-03-07)
Yuichiro Kezuka, Manabu Ohishi, Yoshikane Itoh, Jun Watanabe, Masaru Mitsutomi, Takeshi Watanabe, Takamasa Nonaka
ABSTRACT
Chitinase C (ChiC) from Streptomyces griseus HUT6037 was the first glycoside hydrolase family 19 chitinase that was found in an organism other than higher plants. An N-terminal chitin-binding domain and a C-terminal catalytic domain connected by a linker peptide constitute ChiC. We determined the crystal structure of full-length ChiC, which is the only representative of the two-domain chitinases in the family. The catalytic domain has an alpha-helix-rich fold with a deep cleft containing a catalytic site, and lacks three loops on the domain surface compared with the catalytic domain of plant chitinases. The chitin-binding domain is an all-beta protein with two tryptophan residues (Trp59 and Trp60) aligned on the surface. We suggest the binding mechanism of tri-N-acetylchitotriose onto the chitin-binding domain on the basis of molecular dynamics (MD) simulations. In this mechanism, the ligand molecule binds well on the surface-exposed binding site through two stacking interactions and two hydrogen bonds and only Trp59 and Trp60 are involved in the binding. Furthermore, the flexibility of the Trp60 side-chain, which may be involved in adjusting the binding surface to fit the surface of crystalline chitin by the rotation of chi2 angle, is shown.
Sigma-Aldrich
Chitinase from Streptomyces griseus, chromatographically purified, lyophilized powder, free of DNA contaminants, suitable for Microbiome research
Sigma-Aldrich
Chitinase from Trichoderma viride, lyophilized powder, ≥600 units/g solid
Sigma-Aldrich
N,N′,N′′-Triacetylchitotriose, ≥93% (HPLC)