Merck
  • TBADH activity in water-miscible organic solvents: correlations between enzyme performance, enantioselectivity and protein structure through spectroscopic studies.
TBADH activity in water-miscible organic solvents: correlations between enzyme performance, enantioselectivity and protein structure through spectroscopic studies.
Organic & biomolecular chemistry (2005-02-26)
Linus Olofsson, Ian A Nicholls, Susanne Wikman
ABSTRACT
The enantioselective reduction of 2-pentanone to (R)- and (S)-2-pentanol by Thermoanaerobacter (formerly Thermoanaerobium) brockii alcohol dehydrogenase (TBADH) in mixtures of water and water-miscible organic solvents was investigated. Significant enzymatic activity was retained in up to 87% methanol, ethanol and acetonitrile. The changes in enzyme activity as a function of organic solvent were correlated to structural alterations of TBADH with a series of spectroscopic studies (fluorescence, fluorescence quenching and circular dichroism (CD)). Interestingly, this study shows that the tetrameric form of TBADH is not critical for catalytic performance.
Sigma-Aldrich
2-Pentanone, ≥98%, FCC, FG
Sigma-Aldrich
2-Pentanone, ultrapure grade, ≥99.5%
Supelco
2-Pentanone, analytical standard
Sigma-Aldrich
2-Pentanone, reagent grade, ≥90%
Sigma-Aldrich
2-Pentanone, suitable for HPLC, 99.5%