Merck
  • Substrate and product specificities of cis-type undecaprenyl pyrophosphate synthase.
Substrate and product specificities of cis-type undecaprenyl pyrophosphate synthase.
The Biochemical journal (2004-09-28)
Annie P-C Chen, Sing-Yang Chang, Yu-Chung Lin, Yang-Sheng Sun, Chao-Tsen Chen, Andrew H-J Wang, Po-Huang Liang
ABSTRACT
UPPS (undecaprenyl pyrophosphate synthase) catalyses consecutive condensation reactions of FPP (farnesyl pyrophosphate) with eight isopentenyl pyrophosphates to generate C55 UPP, which serves as a lipid carrier for bacterial peptidoglycan biosynthesis. We reported the co-crystal structure of Escherichia coli UPPS in complex with FPP. Its phosphate head-group is bound to positively charged arginine residues and the hydrocarbon moiety interacts with hydrophobic amino acids including L85, L88 and F89, located on the alpha3 helix of UPPS. We now show that the monophosphate analogue of FPP binds UPPS with an eight times lower affinity (K(d)=4.4 microM) compared with the pyrophosphate analogue, a result of a larger dissociation rate constant (k(off)=192 s(-1)). Farnesol (1 mM) lacking the pyrophosphate does not inhibit the UPPS reaction. GGPP (geranylgeranyl pyrophosphate) containing a larger C20 hydrocarbon tail is an equally good substrate (K(m)=0.3 microM and kcat=2.1 s(-1)) compared with FPP. The shorter C10 GPP (geranyl pyrophosphate) displays a 90-fold larger K(m) value (36.0+/-0.1 microM) but similar kcat value (1.7+/-0.1 s(-1)) compared with FPP. Replacement of L85, L88 or F89 with Ala increases FPP and GGPP K(m) values by the same amount, indicating that these amino acids are important for substrate binding, but do not determine substrate specificity. With GGPP as a substrate, UPPS still catalyses eight isopentenyl pyrophosphate condensation reactions to synthesize C60 product. Computer modelling suggests that the upper portion of the active-site tunnel, where cis double bonds of the product reside, may be critical for determining the final product chain length.
Product Description
Supelco
Geranyl pyrophosphate lithium salt, analytical standard
Sigma-Aldrich
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)
Sigma-Aldrich
Geranyl pyrophosphate lithium salt, ≥95.0% (TLC)