Merck
  • 2-Amino-2-thiazoline and its 1:1 organic salt with 2-naphthoxyacetic acid.
2-Amino-2-thiazoline and its 1:1 organic salt with 2-naphthoxyacetic acid.
Acta crystallographica. Section C, Crystal structure communications (2004-09-04)
Daniel E Lynch
ABSTRACT
The crystal structures of 2-amino-2-thiazoline, C3H6N2S, and 2-amino-2-thiazolinium 2-naphthoxyacetate, C3H7N2S+.C12H9O3-, are reported. The structure of 2-amino-2-thiazoline consists of two unique molecules that construct a convoluted hydrogen-bonded ribbon involving R(2)2(8) graph-set association via both N-H...N and N-H...S interactions. The organic salt structure consists of the two molecules associated via an R(2)2(8) graph-set dimer through N-H...O interactions, with the hydrogen-bonding network propagated via additional N-H...O three-centre interactions from the second 2-amine H atom.
Sigma-Aldrich
2-Naphthoxyacetic acid, suitable for plant cell culture, BioReagent, crystalline
Sigma-Aldrich
2-Naphthoxyacetic acid, 98%