Merck
  • TRPV1+ neurons alter Staphylococcus aureus skin infection outcomes by affecting macrophage polarization and neutrophil recruitment.
TRPV1+ neurons alter Staphylococcus aureus skin infection outcomes by affecting macrophage polarization and neutrophil recruitment.
BMC immunology (2023-12-22)
Changyu Huang, Yang Chen, Yuanqing Cai, Haiqi Ding, Jiaoying Hong, Shan You, Yiming Lin, Hongxin Hu, Yongfa Chen, Xueni Hu, Yanshu Chen, Ying Huang, Chaofan Zhang, Yunzhi Lin, Zida Huang, Wenbo Li, Wenming Zhang, Xinyu Fang
ABSTRACT
The interaction between the nervous system and the immune system can affect the outcome of a bacterial infection. Staphylococcus aureus skin infection is a common infectious disease, and elucidating the relationship between the nervous system and immune system may help to improve treatment strategies. In this study, we found that the local release of calcitonin gene-related peptide (CGRP) increased during S. aureus skin infection, and S. aureus could promote the release of CGRP from transient receptor potential cation channel subfamily V member 1 (TRPV1+) neurons in vitro. The existence of TRPV1+ neurons inhibited the recruitment of neutrophils to the infected region and regulated the polarization of macrophages toward M2 while inhibiting polarization toward M1. This reduces the level of inflammation in the infected area, which aggravates the local infection. Furthermore, this study demonstrates that TRPV1 may be a target for the treatment of S. aureus skin infections and that botulinum neurotoxin A (BoNT/A) and BIBN4096 may reverse the inhibited inflammatory effect of CGRP, making them potential therapeutics for the treatment of skin infection in S. aureus. In S. aureus skin infection, TRPV1+ neurons inhibit neutrophil recruitment and regulate macrophage polarization by releasing CGRP. BoNT/A and BIBN4096 may be potential therapeutic agents for S. aureus skin infection.
Sigma-Aldrich
Anti-Calcitonin Gene-Related Peptide antibody, Mouse monoclonal, clone 4901, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG (γ-chain specific) antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
Sigma-Aldrich
Anti-CD80 (B7-1) Antibody (mouse), PE, clone 16-10A1, clone 16-10A1, 0.2 mg/mL, from hamster(Armenian)
Sigma-Aldrich
Anti-F4/80 antibody, Rabbit monoclonal, clone SP115, recombinant, expressed in proprietary host, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-MRC1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0387, purified immunoglobulin, buffered aqueous glycerol solution