Merck
  • Backbone-Photodegradable Polymers by Incorporating Acylsilane Monomers via Ring-Opening Metathesis Polymerization.
Backbone-Photodegradable Polymers by Incorporating Acylsilane Monomers via Ring-Opening Metathesis Polymerization.
Journal of the American Chemical Society (2021-10-23)
Banruo Huang, Mufeng Wei, Emma Vargo, Yiwen Qian, Ting Xu, F Dean Toste
ABSTRACT
Materials capable of degradation upon exposure to light hold promise in a diverse range of applications including biomedical devices and smart coatings. Despite the rapid access to macromolecules with diverse compositions and architectures enabled by ring-opening metathesis polymerization (ROMP), a general strategy to introduce facile photodegradability into these polymers is lacking. Here, we report copolymers synthesized via ROMP that can be degraded by cleaving the backbone in both solution and solid states under irradiation with a 52 W, 390 nm Kessil LED to generate heterotelechelic low-molecular-weight fragments. To the best of our knowledge, this work represents the first instance of the incorporation of acylsilanes into a polymer backbone. Mechanistic investigation of the degradation process supports the intermediacy of an α-siloxy carbene, formed via a 1,2-photo Brook rearrangement, which undergoes insertion into water followed by cleavage of the resulting hemiacetal.
Sigma-Aldrich
Silyl-ether based ROMP monomer, iPrSi
Sigma-Aldrich
Silyl-ether based ROMP monomer, EtSi
Sigma-Aldrich
Silyl-ether based ROMP Monomer, PhSi