Merck
  • Transcriptional CDK Inhibitors as Potential Treatment Option for Testicular Germ Cell Tumors.
Transcriptional CDK Inhibitors as Potential Treatment Option for Testicular Germ Cell Tumors.
Cancers (2022-04-13)
Kai Funke, Robert Düster, Prince De-Graft Wilson, Lena Arévalo, Matthias Geyer, Hubert Schorle
ABSTRACT
Type II testicular germ cell tumors (TGCT) are the most frequently diagnosed solid malignancy in young men. Up to 15% of patients with metastatic non-seminomas show cisplatin resistance and a very poor survival rate due to lacking treatment options. Transcriptional cyclin-dependent kinases (CDK) have been shown to be effective targets in the treatment of different types of cancer. Here, we investigated the effects of the CDK inhibitors dinaciclib, flavopiridol, YKL-5-124, THZ1, NVP2, SY0351 and THZ531. An XTT viability assay revealed a strong cytotoxic impact of CDK7/12/13 inhibitor SY0351 and CDK9 inhibitor NVP2 on the TGCT wild-type cell lines (2102EP, NCCIT, TCam2) and the cisplatin-resistant cell lines (2102EP-R, NCCIT-R). The CDK7 inhibitor YKL-5-124 showed a strong impact on 2102EP, 2102EP-R, NCCIT and NCCIT-R cell lines, leaving the MPAF control cell line mostly unaffected. FACS-based analysis revealed mild effects on the cell cycle of 2102EP and TCam2 cells after SY0351, YKL-5-124 or NVP2 treatment. Molecular analysis showed a cell-line-specific response for SY0351 and NVP2 inhibition while YKL-5-124 induced similar molecular changes in 2102EP, TCam2 and MPAF cells. Thus, after TGCT subtype determination, CDK inhibitors might be a potential alternative for optimized and individualized therapy independent of chemotherapy sensitivity.
Sigma-Aldrich
Anti-CDK12 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid