Merck
  • Phylogenetic Articulation of Uric Acid Evolution in Mammals and How It Informs a Therapeutic Uricase.
Phylogenetic Articulation of Uric Acid Evolution in Mammals and How It Informs a Therapeutic Uricase.
Molecular biology and evolution (2021-11-01)
Ze Li, Yosuke Hoshino, Lily Tran, Eric A Gaucher
ABSTRACT
The role of uric acid during primate evolution has remained elusive ever since it was discovered over 100 years ago that humans have unusually high levels of the small molecule in our serum. It has been difficult to generate a neutral or adaptive explanation in part because the uricase enzyme evolved to become a pseudogene in apes thus masking typical signals of sequence evolution. Adding to the difficulty is a lack of clarity on the functional role of uric acid in apes. One popular hypothesis proposes that uric acid is a potent antioxidant that increased in concentration to compensate for the lack of vitamin C synthesis in primate species ∼65 Ma. Here, we have expanded on our previous work with resurrected ancient uricase proteins to better resolve the reshaping of uricase enzymatic activity prior to ape evolution. Our results suggest that the pivotal death-knell to uricase activity occurred between 20 and 30 Ma despite small sequential modifications to its catalytic efficiency for the tens of millions of years since primates lost their ability to synthesize vitamin C, and thus the two appear uncorrelated. We also use this opportunity to demonstrate how molecular evolution can contribute to biomedicine by presenting ancient uricases to human immune cells that assay for innate reactivity against foreign antigens. A highly stable and highly catalytic ancient uricase is shown to elicit a lower immune response in more human haplotypes than other uricases currently in therapeutic development.
Sigma-Aldrich
Uricase from Candida sp., recombinant, expressed in E. coli, lyophilized powder, ≥2 units/mg solid
Sigma-Aldrich
Pancreatin from porcine pancreas, 8 × USP specifications